Assay Details
Gene Symbol: Cntfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003929.1 | NP_001003929.1 | 4 - 5 | 521 | 76 | |
XM_006238055.2 | XP_006238117.1 | 3 - 4 | 443 | 76 | ||
GenBank mRNA | S54212.1 | - | 4 - 5 | 521 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
