Assay Details
Gene Symbol: Hivep2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024137.1 | NP_077051.1 | 4 - 5 | 6077 | 116 | |
GenBank mRNA | D37951.1 | - | 4 - 5 | 6077 | 116 | |
M65251.1 | - | 2 - 3 | 750 | 116 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
