Assay Details
Gene Symbol: Bclaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047852.2 | NP_001041317.2 | 10 - 11 | 2630 | 66 | |
XM_006227652.2 | XP_006227714.1 | 11 - 12 | 2797 | 66 | ||
XM_006227653.2 | XP_006227715.1 | 9 - 10 | 2626 | 66 | ||
XM_006227654.2 | XP_006227716.1 | 10 - 11 | 2654 | 66 | ||
XM_008758605.1 | XP_008756827.1 | 10 - 11 | 3789 | 66 | ||
GenBank mRNA | BC161968.1 | - | 10 - 11 | 2630 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of apoptotic process
regulation of DNA-templated transcription in response to stress
negative regulation of transcription, DNA-templated
positive regulation of DNA-templated transcription, initiation
positive regulation of response to DNA damage stimulus
positive regulation of intrinsic apoptotic signaling pathway
DNA binding
RNA binding
positive regulation of apoptotic process
regulation of DNA-templated transcription in response to stress
negative regulation of transcription, DNA-templated
positive regulation of DNA-templated transcription, initiation
positive regulation of response to DNA damage stimulus
positive regulation of intrinsic apoptotic signaling pathway
DNA binding
RNA binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P158537 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 405603 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 945670 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 205275 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 533773 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 140280 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 761169 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 240221 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 678576 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 889954 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 860958 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 126354 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 241092 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 895191 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 398449 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 571599 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 292581 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 968638 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 706689 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 155704 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 157560 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P158537 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P158537 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P158537 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P278562 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P356236 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10702000 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702008 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702005 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702006 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10701999 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702003 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702007 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702013 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10701994 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10701996 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702012 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702014 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10701997 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10701998 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702001 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10701990 | - | - |
Rat Genome 230 2.0 Array | 1383601_at | - | - |
Rat Genome 230 2.0 Array | 1388752_at | - | - |
Rat Genome 230 2.0 Array | 1388752_PM_at | - | - |
Rat Genome 230 2.0 Array | 1373868_PM_at | - | - |
Rat Genome 230 2.0 Array | 1394359_PM_at | - | - |
Rat Genome 230 2.0 Array | 1373868_at | - | - |
Rat Genome 230 2.0 Array | 1383601_PM_at | - | - |
Rat Genome 230 2.0 Array | 1394359_at | - | - |
Affymetrix HT Rat Focus Array | 1394359_at | - | - |
Affymetrix HT Rat Focus Array | 1373868_at | - | - |
Affymetrix HT Rat Focus Array | 1383601_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1394359_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1388752_at | - | - |
Affymetrix HT Rat Focus Array | 1373868_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1388752_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1383601_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0100064616.rn.1 | TC0100008002.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100064585.rn.1 | TC0100008002.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100064591.rn.1 | TC0100008002.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100064595.rn.1 | TC0100008002.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100064604.rn.1 | TC0100008002.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100064594.rn.1 | TC0100008002.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100064602.rn.1 | TC0100008002.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100064589.rn.1 | TC0100008002.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100064587.rn.1 | TC0100008002.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100064586.rn.1 | TC0100008002.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100064590.rn.1 | TC0100008002.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100064596.rn.1 | TC0100008002.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100064597.rn.1 | TC0100008002.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100064599.rn.1 | TC0100008002.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100064605.rn.1 | TC0100008002.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100064614.rn.1 | TC0100008002.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100064615.rn.1 | TC0100008002.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100064598.rn.1 | TC0100008002.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100064606.rn.1 | TC0100008002.rn.1 | RTA TC |
Rat Expression Set 230 | 1388752_PM_at | - | - |
Rat Expression Set 230 | 1388752_at | - | - |
Rat Expression Set 230 | 1373868_PM_at | - | - |
Rat Expression Set 230 | 1373868_at | - | - |
Rat Expression Set 230 | 1383601_PM_at | - | - |
Rat Expression Set 230 | 1383601_at | - | - |
Rat Expression Set 230 | 1394359_at | - | - |
Rat Expression Set 230 | 1394359_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10701990 | - | - |
Rat Genome U34 Set | rc_AA800673_at | - | - |
Rat Genome U34 Set | rc_AA926089_at | - | - |
Rat Genome U34 Set | rc_AI136833_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1355583 | - | - |
