Assay Details
Gene Symbol: Bclaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047852.2 | NP_001041317.2 | 11 - 12 | 2777 | 110 | |
XM_006227652.2 | XP_006227714.1 | 12 - 13 | 2944 | 110 | ||
XM_006227653.2 | XP_006227715.1 | 10 - 11 | 2773 | 110 | ||
XM_006227654.2 | XP_006227716.1 | 11 - 12 | 2801 | 110 | ||
XM_008758605.1 | XP_008756827.1 | 11 - 12 | 3936 | 110 | ||
GenBank mRNA | BC161968.1 | - | 11 - 12 | 2777 | 110 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
