Assay Details
Gene Symbol: Aldh8a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191088.1 | NP_001178017.1 | 1 - 2 | 174 | 156 | |
XM_006227674.2 | XP_006227736.1 | 1 - 2 | 209 | 156 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
