Assay Details
Gene Symbol: Lats1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134543.2 | NP_001128015.2 | 1 - 2 | 226 | 72 | |
GenBank mRNA | FN804841.1 | - | 1 - 2 | 226 | 72 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001134543.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
