Assay Details
Gene Symbol: Lats1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134543.2 | NP_001128015.2 | 3 - 4 | 855 | 106 | |
XM_006227614.2 | XP_006227676.1 | 2 - 3 | 1497 | 106 | ||
XM_008758607.1 | XP_008756829.1 | 2 - 3 | 1497 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
