Assay Details
Gene Symbol: Lats1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134543.2 | NP_001128015.2 | 6 - 7 | 3129 | 111 | |
XM_006227614.2 | XP_006227676.1 | 5 - 6 | 3771 | 111 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
