Assay Details
Gene Symbol: Sim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107641.1 | NP_001101111.1 | 1 - 2 | 611 | 105 | |
XM_006256604.2 | XP_006256666.1 | 2 - 3 | 899 | 105 | ||
XM_008772994.1 | XP_008771216.1 | 2 - 3 | 739 | 105 | ||
XM_008772995.1 | XP_008771217.1 | 2 - 3 | 2088 | 105 | ||
XM_008772996.1 | XP_008771218.1 | 2 - 3 | 722 | 105 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
