Assay Details
Gene Symbol: Sim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107641.1 | NP_001101111.1 | 4 - 5 | 894 | 105 | |
XM_006256604.2 | XP_006256666.1 | 5 - 6 | 1182 | 105 | ||
XM_008772994.1 | XP_008771216.1 | 5 - 6 | 1022 | 105 | ||
XM_008772995.1 | XP_008771217.1 | 5 - 6 | 2371 | 105 | ||
XM_008772996.1 | XP_008771218.1 | 5 - 6 | 1005 | 105 | ||
XM_008772997.1 | XP_008771219.1 | 4 - 5 | 4647 | 105 | ||
XM_008772998.1 | XP_008771220.1 | 2 - 3 | 179 | 105 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the XM_008772998.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
