Assay Details
Gene Symbol: Sim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107641.1 | NP_001101111.1 | 7 - 8 | 1282 | 144 | |
XM_006256604.2 | XP_006256666.1 | 8 - 9 | 1570 | 144 | ||
XM_008772994.1 | XP_008771216.1 | 8 - 9 | 1410 | 144 | ||
XM_008772995.1 | XP_008771217.1 | 8 - 9 | 2759 | 144 | ||
XM_008772996.1 | XP_008771218.1 | 8 - 9 | 1393 | 144 | ||
XM_008772997.1 | XP_008771219.1 | 7 - 8 | 5035 | 144 | ||
XM_008772998.1 | XP_008771220.1 | 5 - 6 | 567 | 144 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
