Assay Details
Gene Symbol: Scml4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107638.1 | NP_001101108.1 | 7 - 8 | 1201 | 96 | |
XM_006256572.2 | XP_006256634.1 | 7 - 8 | 1498 | 96 | ||
XM_008772989.1 | XP_008771211.1 | 3 - 4 | 1382 | 96 | ||
XM_008772990.1 | XP_008771212.1 | 3 - 4 | 798 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P017766 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 546245 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 357470 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 374190 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 211092 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 155314 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 951933 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 642099 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 829547 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 811946 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 204496 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 5613 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 386420 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427768 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225835 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 921354 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 333431 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 836285 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 709362 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 280090 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 237216 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 796440 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 560205 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 971807 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 535269 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 394750 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 420740 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 82677 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 577349 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 303063 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 914398 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 188184 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 237597 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 221745 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 713495 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 424891 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 320815 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 580471 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 863095 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 800212 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 779677 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 809017 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 218099 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 388765 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 863590 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 780428 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 497619 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 434469 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 144990 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 837231 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 109551 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 327528 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 518228 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 464440 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 742279 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 205181 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 810478 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 214278 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763351 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244649 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 758769 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 865827 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 610550 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 124968 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 899426 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 367660 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 916321 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 723765 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 78230 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600066 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 221460 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 400722 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 582443 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 33030 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 32305 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 26990 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 160863 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 4613 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 123373 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 74759 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 155880 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 89124 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 101836 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 55950 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 88559 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P017766 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P017766 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P017766 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P734395 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 14962 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 15949 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 370 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10830609 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10830611 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10830612 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10830610 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10830607 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10830606 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10830605 | - | - |
Affymetrix Rat Clariom S Assay | PSR2000007161.rn.1 | TC2000000849.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR2000007160.rn.1 | TC2000000849.rn.1 | RTA TC |
Affymetrix Rat Gene 1.0 ST Array | 10830605 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21389190 | - | - |
ABI Rat Genome Survey Microarray | 21861443 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1362365 | - | - |
