Assay Details
Gene Symbol: Il1r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013123.3 | NP_037255.3 | 1 - 2 | 43 | 121 | |
XM_006244746.2 | XP_006244808.1 | 2 - 3 | 2152 | 121 | ||
XM_006244747.2 | XP_006244809.1 | 2 - 3 | 409 | 121 | ||
GenBank mRNA | AB112437.1 | - | 1 - 2 | 43 | 121 | |
M95578.1 | - | 2 - 3 | 364 | 121 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
