Assay Details
Gene Symbol: Cnga3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053495.1 | NP_445947.1 | 1 - 2 | 56 | 81 | |
GenBank mRNA | AB002801.1 | - | 1 - 2 | 56 | 81 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AB002801.1, NM_053495.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
