Assay Details
Gene Symbol: Chst10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006244730.2 | XP_006244792.1 | 1 - 2 | 264 | 87 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006244730.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
