Assay Details
Gene Symbol: Hspd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022229.2 | NP_071565.2 | 7 - 8 | 926 | 64 | |
XM_006244932.2 | XP_006244994.1 | 7 - 8 | 962 | 64 | ||
GenBank mRNA | BC086507.1 | - | 7 - 8 | 926 | 64 | |
X53585.1 | - | 6 - 7 | 790 | 64 | ||
X54793.1 | - | 6 - 7 | 874 | 64 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
