Assay Details
Gene Symbol: Ddx41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108046.2 | NP_001101516.1 | 9 - 10 | 941 | 97 | |
GenBank mRNA | BC166825.1 | - | 9 - 10 | 941 | 97 |
Gene Symbol: LOC100910720 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008771481.1 | XP_008769703.1 | 786 | 97 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
