Assay Details
Gene Symbol: Grk6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112712.1 | NP_001106183.1 | 12 - 13 | 1272 | 73 | |
NM_001112713.1 | NP_001106184.1 | 12 - 13 | 1272 | 73 | ||
NM_031657.3 | NP_113845.2 | 12 - 13 | 1272 | 73 | ||
XM_006253653.2 | XP_006253715.1 | 12 - 13 | 1381 | 73 | ||
XM_008771504.1 | XP_008769726.1 | 11 - 12 | 1539 | 73 | ||
GenBank mRNA | AF040750.1 | - | 12 - 13 | 1272 | 73 | |
X97439.1 | - | 12 - 13 | 1272 | 73 | ||
X97440.1 | - | 12 - 13 | 1272 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
