Assay Details
Gene Symbol: Rnf44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024795.1 | NP_001019966.1 | 7 - 8 | 1135 | 67 | |
XM_006253633.2 | XP_006253695.1 | 7 - 8 | 2132 | 67 | ||
XM_006253634.2 | XP_006253696.1 | 7 - 8 | 1125 | 67 | ||
XM_006253637.2 | XP_006253699.1 | 7 - 8 | 2654 | 67 | ||
XM_006253641.2 | XP_006253703.1 | 7 - 8 | 2068 | 67 | ||
XM_006253642.2 | XP_006253704.1 | 7 - 8 | 2590 | 67 | ||
XM_006253644.2 | XP_006253706.1 | 7 - 8 | 1061 | 67 | ||
XM_008771498.1 | XP_008769720.1 | 7 - 8 | 2651 | 67 | ||
GenBank mRNA | BC098030.1 | - | 7 - 8 | 1135 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
