Assay Details
Gene Symbol: Fastk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006235892.2 | XP_006235954.1 | 9 - 10 | 1635 | 91 | |
XM_006235894.2 | XP_006235956.1 | 9 - 10 | 1622 | 91 | ||
XM_006235895.1 | XP_006235957.1 | 9 - 10 | 1619 | 91 | ||
GenBank mRNA | FQ215763.1 | - | 10 - 11 | 1497 | 91 | |
FQ228972.1 | - | 9 - 10 | 1576 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
