Assay Details
Gene Symbol: Fastk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006235892.2 | XP_006235954.1 | 8 - 9 | 1522 | 96 | |
XM_006235894.2 | XP_006235956.1 | 8 - 9 | 1509 | 96 | ||
XM_006235895.1 | XP_006235957.1 | 8 - 9 | 1506 | 96 | ||
GenBank mRNA | FQ215763.1 | - | 9 - 10 | 1384 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
