Assay Details
Gene Symbol: Asic3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173135.1 | NP_775158.1 | 10 - 11 | 1522 | 71 | |
GenBank mRNA | AF013598.1 | - | 10 - 11 | 1522 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
