Assay Details
Gene Symbol: Abcf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109666.1 | NP_001103136.1 | 12 - 13 | 1711 | 98 | |
XM_006235929.2 | XP_006235991.1 | 13 - 14 | 1681 | 98 | ||
XM_006235930.2 | XP_006235992.1 | 13 - 14 | 1843 | 98 | ||
XM_006235931.2 | XP_006235993.1 | 13 - 14 | 1678 | 98 | ||
XM_006235932.2 | XP_006235994.1 | 13 - 14 | 1672 | 98 | ||
XM_006235933.2 | XP_006235995.1 | 13 - 14 | 1669 | 98 | ||
XM_006235934.2 | XP_006235996.1 | 12 - 13 | 1591 | 98 | ||
GenBank mRNA | BC129119.1 | - | 12 - 13 | 1578 | 98 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
