Assay Details
Gene Symbol: Pmpcb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022395.2 | NP_071790.1 | 5 - 6 | 666 | 130 | |
GenBank mRNA | BC078826.1 | - | 5 - 6 | 661 | 130 | |
D13907.1 | - | 5 - 6 | 644 | 130 | ||
L12965.1 | - | 5 - 6 | 667 | 130 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
