Assay Details
Gene Symbol: Pmpcb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022395.2 | NP_071790.1 | 8 - 9 | 1001 | 89 | |
GenBank mRNA | BC078826.1 | - | 8 - 9 | 996 | 89 | |
D13907.1 | - | 8 - 9 | 979 | 89 | ||
L12965.1 | - | 8 - 9 | 1002 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
