Assay Details
Gene Symbol: Psmc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033236.1 | NP_150239.1 | 7 - 8 | 611 | 63 | |
GenBank mRNA | BC061542.1 | - | 7 - 8 | 653 | 63 | |
D50694.1 | - | 7 - 8 | 611 | 63 | ||
FQ209873.1 | - | 4 - 5 | 414 | 63 | ||
FQ212821.1 | - | 7 - 8 | 641 | 63 | ||
FQ213265.1 | - | 4 - 5 | 414 | 63 | ||
FQ215767.1 | - | 4 - 5 | 414 | 63 | ||
FQ225383.1 | - | 4 - 5 | 414 | 63 | ||
FQ234824.1 | - | 4 - 5 | 414 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
