Assay Details
Gene Symbol: Kcnh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053949.1 | NP_446401.1 | 9 - 10 | 2401 | 102 | |
XM_006235867.2 | XP_006235929.1 | 9 - 10 | 2534 | 102 | ||
XM_008762630.1 | XP_008760852.1 | 5 - 6 | 1722 | 102 | ||
GenBank mRNA | AY669863.1 | - | 5 - 6 | 1375 | 102 | |
Z96106.1 | - | 9 - 10 | 2401 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
