Assay Details
Gene Symbol: Mrpl15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006237805.2 | XP_006237867.1 | 4 - 5 | 602 | 78 | |
GenBank mRNA | FQ214529.1 | - | 4 - 5 | 564 | 78 | |
FQ216137.1 | - | 4 - 5 | 543 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
