Assay Details
Gene Symbol: Sdcbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031986.1 | NP_114192.1 | 5 - 6 | 477 | 106 | |
GenBank mRNA | AF248548.1 | - | 5 - 6 | 477 | 106 | |
AJ292243.1 | - | 4 - 5 | 420 | 106 | ||
BC064651.1 | - | 5 - 6 | 445 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
