Assay Details
Gene Symbol: Sec14l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053801.2 | NP_446253.2 | 7 - 8 | 622 | 89 | |
XM_006251154.2 | XP_006251216.1 | 7 - 8 | 611 | 89 | ||
GenBank mRNA | AF309558.1 | - | 7 - 8 | 579 | 89 | |
BC089785.1 | - | 7 - 8 | 622 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
