Assay Details
Gene Symbol: Polm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011912.1 | NP_001011912.1 | 1 - 2 | 449 | 129 | |
XM_006251213.2 | XP_006251275.1 | 1 - 2 | 818 | 129 | ||
GenBank mRNA | BC081868.1 | - | 1 - 2 | 449 | 129 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
