Assay Details
Gene Symbol: Polm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011912.1 | NP_001011912.1 | 9 - 10 | 1568 | 70 | |
XM_006251211.2 | XP_006251273.1 | 9 - 10 | 1783 | 70 | ||
XM_006251212.2 | XP_006251274.1 | 9 - 10 | 1771 | 70 | ||
XM_006251213.2 | XP_006251275.1 | 9 - 10 | 1949 | 70 | ||
XM_006251214.2 | XP_006251276.1 | 9 - 10 | 1191 | 70 | ||
GenBank mRNA | BC081868.1 | - | 9 - 10 | 1568 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
