Assay Details
Gene Symbol: Mfsd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191680.1 | NP_001178609.1 | 8 - 9 | 1023 | 98 | |
XM_006251243.2 | XP_006251305.1 | 9 - 10 | 1325 | 98 | ||
XM_006251244.2 | XP_006251306.1 | 9 - 10 | 1322 | 98 | ||
XM_006251245.2 | XP_006251307.1 | 9 - 10 | 1158 | 98 | ||
XM_006251246.2 | XP_006251308.1 | 9 - 10 | 1155 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
