Assay Details
Gene Symbol: Uvssa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134558.1 | NP_001128030.1 | 13 - 14 | 2521 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P072068 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 235750 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 375908 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 90039 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 557251 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 72260 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 560566 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 910958 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 674643 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 860860 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 882463 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 836947 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 350332 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 953061 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 537918 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 94154 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 920427 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 112773 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 563226 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 152839 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 223405 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 612506 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667694 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912423 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 49822 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 138854 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 43349 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 849783 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 419168 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 386445 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 796235 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 258181 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 75206 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 79883 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 107053 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 84303 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P072068 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P072068 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P072068 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P445465 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10773624 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10773627 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10773626 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10773631 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10773634 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10773635 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10773629 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10773623 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10773630 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10773625 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10773632 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10773633 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10773628 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10773622 | - | - |
Affymetrix Rat Clariom S Assay | PSR1400015333.rn.1 | TC1400002111.rn.1 | RTA TC |
Affymetrix Rat Gene 1.0 ST Array | 10773622 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22367356 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1372361 | - | - |
