Assay Details
Gene Symbol: Pold2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013050.1 | NP_001013068.1 | 10 - 11 | 1320 | 101 | |
GenBank mRNA | BC079267.1 | - | 10 - 11 | 1320 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
