Assay Details
Gene Symbol: Pold2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013050.1 | NP_001013068.1 | 8 - 9 | 1100 | 63 | |
GenBank mRNA | BC079267.1 | - | 8 - 9 | 1100 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
