Assay Details
Gene Symbol: Tmed4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006251444.2 | XP_006251506.1 | 1 - 2 | 212 | 62 | |
XM_006251445.2 | XP_006251507.1 | 1 - 2 | 213 | 62 | ||
GenBank mRNA | BC168726.1 | - | 1 - 2 | 184 | 62 | |
BC168735.1 | - | 1 - 2 | 184 | 62 | ||
BC168766.1 | - | 1 - 2 | 184 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
