Assay Details
Gene Symbol: Anxa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012823.2 | NP_036955.2 | 4 - 5 | 341 | 100 | |
XM_006250694.2 | XP_006250756.1 | 4 - 5 | 705 | 100 | ||
XM_006250696.2 | XP_006250758.1 | 5 - 6 | 461 | 100 | ||
GenBank mRNA | BC081856.1 | - | 3 - 4 | 253 | 100 | |
FQ232970.1 | - | 4 - 5 | 332 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
