Assay Details
Gene Symbol: Arhgap24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012032.1 | NP_001012032.1 | 2 - 3 | 580 | 68 | |
XM_006250652.2 | XP_006250714.1 | 3 - 4 | 786 | 68 | ||
XM_006250654.2 | XP_006250716.1 | 3 - 4 | 3368 | 68 | ||
XM_008770017.1 | XP_008768239.1 | 3 - 4 | 1447 | 68 | ||
XM_008770018.1 | XP_008768240.1 | 2 - 3 | 1299 | 68 | ||
GenBank mRNA | AB108670.1 | - | 2 - 3 | 782 | 68 | |
BC085797.1 | - | 2 - 3 | 580 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
