Assay Details
Gene Symbol: Arhgap24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012032.1 | NP_001012032.1 | 3 - 4 | 717 | 100 | |
XM_006250652.2 | XP_006250714.1 | 4 - 5 | 923 | 100 | ||
XM_006250654.2 | XP_006250716.1 | 4 - 5 | 3505 | 100 | ||
XM_008770017.1 | XP_008768239.1 | 4 - 5 | 1584 | 100 | ||
XM_008770018.1 | XP_008768240.1 | 3 - 4 | 1436 | 100 | ||
GenBank mRNA | AB108670.1 | - | 3 - 4 | 919 | 100 | |
BC085797.1 | - | 3 - 4 | 717 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
