Assay Details
Gene Symbol: Mrpl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105997.2 | NP_001099467.1 | 3 - 4 | 421 | 169 | |
XM_008770009.1 | XP_008768231.1 | 4 - 5 | 788 | 169 | ||
GenBank mRNA | BC168772.1 | - | 3 - 4 | 421 | 169 | |
FQ217255.1 | - | 3 - 4 | 423 | 169 |
Gene Symbol: LOC100359687 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750509.3 | XP_003750557.1 | 3 - 4 | 415 | 169 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
