Assay Details
Gene Symbol: Mrpl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105997.2 | NP_001099467.1 | 6 - 7 | 689 | 162 | |
XM_008770009.1 | XP_008768231.1 | 7 - 8 | 1056 | 162 | ||
GenBank mRNA | BC168772.1 | - | 6 - 7 | 689 | 162 | |
FQ217255.1 | - | 6 - 7 | 691 | 162 |
Gene Symbol: LOC100359687 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750509.3 | XP_003750557.1 | 6 - 7 | 683 | 162 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
