Assay Details
Gene Symbol: Hnrnpdl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033696.1 | NP_001028868.1 | 6 - 7 | 1067 | 102 | |
XM_006250662.2 | XP_006250724.1 | 6 - 7 | 1203 | 102 | ||
XM_006250663.2 | XP_006250725.1 | 6 - 7 | 1203 | 102 | ||
XM_008770024.1 | XP_008768246.1 | 6 - 7 | 1203 | 102 | ||
GenBank mRNA | BC104683.1 | - | 6 - 7 | 1067 | 102 | |
FQ222460.1 | - | 1797 | 102 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
