Assay Details
Gene Symbol: Pcyt1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078622.2 | NP_511177.2 | 3 - 4 | 320 | 132 | |
XM_006248444.2 | XP_006248506.1 | 3 - 4 | 473 | 132 | ||
XM_008768694.1 | XP_008766916.1 | 3 - 4 | 671 | 132 | ||
GenBank mRNA | AW919077.1 | - | 3 - 4 | 321 | 132 | |
BC085713.1 | - | 3 - 4 | 318 | 132 | ||
L13245.1 | - | 2 - 3 | 217 | 132 | ||
M36071.1 | - | 3 - 4 | 317 | 132 | ||
U03490.1 | - | 3 - 4 | 282 | 132 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
