Assay Details
Gene Symbol: Pcyt1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078622.2 | NP_511177.2 | 1 - 2 | 87 | 109 | |
GenBank mRNA | AW919077.1 | - | 1 - 2 | 88 | 109 | |
BC085713.1 | - | 1 - 2 | 85 | 109 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC085713.1, NM_078622.2 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
