Assay Details
Gene Symbol: Cldnd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006955.1 | NP_001006956.1 | 1 - 2 | 90 | 75 | |
XM_006248205.2 | XP_006248267.1 | 1 - 2 | 231 | 75 | ||
GenBank mRNA | BC083649.1 | - | 1 - 2 | 90 | 75 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC083649.1, NM_001006955.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
