Assay Details
Gene Symbol: Cldnd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006955.1 | NP_001006956.1 | 2 - 3 | 395 | 114 | |
XM_006248204.2 | XP_006248266.1 | 3 - 4 | 587 | 114 | ||
XM_006248205.2 | XP_006248267.1 | 2 - 3 | 536 | 114 | ||
XM_006248206.2 | XP_006248268.1 | 3 - 4 | 438 | 114 | ||
XM_008768633.1 | XP_008766855.1 | 2 - 3 | 378 | 114 | ||
GenBank mRNA | BC083649.1 | - | 2 - 3 | 395 | 114 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
