Assay Details
Gene Symbol: Gsk3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032080.1 | NP_114469.1 | 7 - 8 | 956 | 131 | |
GenBank mRNA | FQ231148.1 | - | 2 - 3 | 224 | 131 | |
FQ233111.1 | - | 2 - 3 | 224 | 131 | ||
X53428.1 | - | 7 - 8 | 931 | 131 | ||
X73653.1 | - | 7 - 8 | 956 | 131 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
