Assay Details
Gene Symbol: Popdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199113.1 | NP_954544.1 | 1 - 2 | 919 | 121 | |
XM_008768733.1 | XP_008766955.1 | 1 - 2 | 2926 | 121 | ||
XM_008768734.1 | XP_008766956.1 | 1 - 2 | 2926 | 121 | ||
GenBank mRNA | AY490241.1 | - | 1 - 2 | 882 | 121 | |
BC061876.1 | - | 1 - 2 | 919 | 121 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
