Assay Details
Gene Symbol: Nr1i2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_052980.2 | NP_443212.1 | 7 - 8 | 1284 | 79 | |
GenBank mRNA | AF151377.1 | - | 7 - 8 | 1257 | 79 | |
BC091138.1 | - | 6 - 7 | 1378 | 79 | ||
BC107449.1 | - | 5 - 6 | 1159 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
